Learn More
Fisherbrand™ Class 100 Vinyl CR Chair - Med Bench Height with Med Back Products for Science Education
Fisherbrand™ Class 100 Vinyl Clean Room Chairs with Medium Back are height adjustable for 36 in. work surfaces and feature ergonomic adjustments, clean room components, arms, and stationary glides. Antibacterial/Antifungal Grade B vinyl is easily cleaned making ideal for clean room environments.
$471.00
Specifications
Base Material Polished Cast Aluminum
Arm Style 6-way adjustable up and down, in and out, twist toward or away from the user
Foot Type Non-marring bell glides (suitable for both hard and carpeted floors)
Footrest Yes
Footrest Size (English) 20 in.
View More Specs
Products 4
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
S0100121 Products for Science Education
View Documents
Orange
Each for $471.00
Add to cart
S0100123 Products for Science Education
View Documents
Dove
Each for $471.00
Add to cart
S0100134 Products for Science Education
View Documents
Taupe
Each for $471.00
Add to cart
S0100156 Products for Science Education
View Documents
White
Each for $471.00
Add to cart
Description
Description
  • Medium Back
  • Adjustable from 21.5 to 29 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • 6-way adjustable arm rests adjust up and down, in and out, and twist toward and away from the user
  • 20-Inch Chrome adjustable foot ring with internal lock
  • Cast-aluminum base
  • Stationary, non-marring glides suitable for hard floors
  • Antibacterial/antifungal Grade B Vinyl
  • Variety of color options to suit any work space
  • Class 100/ISO 5 Clean Room
Specifications
Specifications
Polished Cast Aluminum
Non-marring bell glides (suitable for both hard and carpeted floors)
20 in.
Cleanroom Seating
45 lb.
No
6-way adjustable up and down, in and out, twist toward or away from the user
Yes
Vinyl
Medium Bench Height
Yes
No