Learn More
Fisherbrand™ Class 100 Vinyl CR Chair - Med Bench Height with Med Back, Seat Tilt, Adj Arms, Foot Ring, and Casters in Grade B Vinyl Products for Science Education
Fisherbrand™ Class 100 Vinyl Clean Room Chairs with Medium Back are height adjustable for 36 in. work surfaces and feature ergonomic adjustments, clean room components, arms, and self-braking casters. Antibacterial/Antifungal Grade B vinyl is easily cleaned making ideal for clean room environments.
$420.00
Specifications
Adjustable Yes
Material Vinyl
Footrest Yes
With Armrest Yes
With Backrest Yes
View More Specs
Products 5
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
S40052 Products for Science Education
View Documents
Blue
Each for $420.00
Add to cart
S40067 Products for Science Education
View Documents
Black
Each for $420.00
Add to cart
S40090 Products for Science Education
View Documents
Burgundy
Each for $420.00
Add to cart
S40108 Products for Science Education
View Documents
Gray
Each for $420.00
Add to cart
S40150 Products for Science Education
View Documents
Dark Blue
Each for $420.00
Add to cart
Description
Description
  • Medium Back
  • Adjustable from 21.5-29 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • 8-degree forward and 3-degree backward locking seat tilt
  • 6-way adjustable arm rests adjust up and down, in and out, and twist toward and away from the user
  • 20-Inch Chrome adjustable foot ring with internal lock
  • Cast-aluminum base
  • Chrome-hooded, dual-wheel, self-braking, non-marring casters suitable for hard floors
  • Antibacterial/antifungal Grade B Vinyl
  • Variety of color options to suit any work space
  • Class 100/ISO 5 Clean Room
Specifications
Specifications
Yes
Yes
Yes
Yes
Vinyl
Yes
Yes
Class 100/ISO Class 5