Learn More
Fisherbrand™ Class 100 Vinyl CR Chair - Med Bench Height with Med Back, Tilt Products for Science Education
Fisherbrand™ Class 100 Vinyl Clean Room Chairs with Medium Back are height adjustable for 36 in. work surfaces and feature ergonomic adjustments, clean room components, arms, and stationary glides. Grade A vinyl, in a variety of colors, offer additional antibacterial properties and a high-end look and feel.
$521.00
Specifications
Base Material Polished Cast Aluminum
Arm Style 6-way adjustable up and down, in and out, twist toward or away from the user
Foot Type Non-marring bell glides (suitable for both hard and carpeted floors)
Footrest Yes
Footrest Size (English) 20 in.
View More Specs
Products 6
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
S0100135 Products for Science Education
View Documents
Marine Blue
Each for $521.00
Add to cart
S0100342 Products for Science Education
View Documents
Borscht
Each for $521.00
Add to cart
S0100356 Products for Science Education
View Documents
Sterling
Each for $521.00
Add to cart
S0100357 Products for Science Education
View Documents
Carbon
Each for $521.00
Add to cart
S0100358 Products for Science Education
View Documents
Storm
Each for $521.00
Add to cart
S0100360 Products for Science Education
View Documents
Taupe
Each for $521.00
Add to cart
Description
Description
  • Medium Back
  • Adjustable from 21.5 to 29 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • 8-degree forward and 3-degree backward locking seat tilt
  • 6-way adjustable arm rests adjust up and down, in and out, and twist toward and away from the user
  • 20-Inch Chrome adjustable foot ring with internal lock
  • Cast-aluminum base
  • Stationary, non-marring glides suitable for hard floors
  • Antibacterial/antifungal Grade A Vinyl
  • Additional antibacterial protections
  • High end look and feel
  • Variety of color options to suit any work space
  • Class 100/ISO Class 5 Clean Room
Specifications
Specifications
Polished Cast Aluminum
Non-marring bell glides (suitable for both hard and carpeted floors)
20 in.
Cleanroom Seating
Medium Bench Height
Yes
Yes
6-way adjustable up and down, in and out, twist toward or away from the user
Yes
Vinyl
Ergonomic 8 degree forward and 3 degree backward seat tilt locks in place
46 lb.
No