Learn More
Fisherbrand™ Class 100 Vinyl CR/ESD Chair - High Bench Height with Med Back, Seat Tilt, Foot Ring, and ESD Stationary Glides in ESD Vinyl Products for Science Education
Fisherbrand™ Class 100 Vinyl Clean Room/ESD Chairs with Medium Back height adjustable for 42 in. work surfaces & feature ergonomic adjustments, clean room & electro-static dissipating components, chrome adjustable foot ring, & stationary glides. Properly grounded for static sensitive applications.
$479.00
Specifications
Adjustable Yes
Footrest Yes
With Armrest No
With Backrest Yes
With Casters No
View More Specs
Products 2
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
S39986 Products for Science Education
View Documents
Black
Each for $479.00
Add to cart
S39988 Products for Science Education
View Documents
Blue
Each for $479.00
Add to cart
Description
Description
  • Medium Back
  • Adjustable from 26.25-36 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • Locking seat tilt
  • Chrome adjustable foot ring
  • Electro-static dissipating materials for proper grounding
  • Cast-aluminum base
  • Stationary ESD glides suitable for hard floors
  • Electro-static dissipating vinyl
  • Meets ANSI/ESD STM12.1-2013
  • Class 100/ISO Class 5 Clean Room
Specifications
Specifications
Yes
No
No
Class 100/ISO Class 5
Yes
Yes
Yes