Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ Class 2 ArcGuard™ Rubber Voltage Glove Kit

National Safety Apparel™ Class 2 ArcGuard™ Rubber Voltage Glove Kit includes leather gloves, class 2 rubber voltage gloves, and a glove bag.

$408.50

Specifications

Certifications/Compliance ASTM CLASS 2
Material Leather, Rubber
For Use With (Application) Voltage, electrical, arc flash
Length (English) 12 in.
Length (Metric) 30.48 cm
View More Specs

Products 15
Catalog Number Mfr. No. Color Size Price Quantity & Availability  
Catalog Number Mfr. No. Color Size Price Quantity & Availability  
17-355-030
View Documents
National Safety Apparel™
KITGC2B08
Black 8 Pair for $408.50
Add to cart
 
17-355-031
View Documents
National Safety Apparel™
KITGC2B09
Black 9 Pair for $408.50
Add to cart
 
17-355-032
View Documents
National Safety Apparel™
KITGC2B10
Black 10 Pair for $408.50
Add to cart
 
17-355-033
View Documents
National Safety Apparel™
KITGC2B11
Black 11 Pair for $408.50
Add to cart
 
17-355-034
View Documents
National Safety Apparel™
KITGC2B12
Black 12 Pair for $408.50
Add to cart
 
17-355-035
View Documents
National Safety Apparel™
KITGC2R08
Red 8 Pair for $408.50
Add to cart
 
17-355-036
View Documents
National Safety Apparel™
KITGC2R09
Red 9 Pair for $408.50
Add to cart
 
17-355-037
View Documents
National Safety Apparel™
KITGC2R10
Red 10 Pair for $408.50
Add to cart
 
17-355-038
View Documents
National Safety Apparel™
KITGC2R11
Red 11 Pair for $408.50
Add to cart
 
17-355-039
View Documents
National Safety Apparel™
KITGC2R12
Red 12 Pair for $408.50
Add to cart
 
17-355-040
View Documents
National Safety Apparel™
KITGC2Y08
Yellow 8 Pair for $408.50
Add to cart
 
17-355-041
View Documents
National Safety Apparel™
KITGC2Y09
Yellow 9 Pair for $408.50
Add to cart
 
17-355-042
View Documents
National Safety Apparel™
KITGC2Y10
Yellow 10 Pair for $408.50
Add to cart
 
17-355-043
View Documents
National Safety Apparel™
KITGC2Y11
Yellow 11 Pair for $408.50
Add to cart
 
17-355-044
View Documents
National Safety Apparel™
KKITGC2Y12
Yellow 12 Pair for $408.50
Add to cart
 
Description

Description

  • Fabric/Material: Rubber, Leather
  • Rubber voltage gloves are electrically tested and marked with initial test date
  • Class 2 - Max Use Voltage 17000 V AC, 25500 V DC
  • Includes: One pair of rubber voltage gloves, 12 in. leather protectors, glove bag
Specifications

Specifications

ASTM CLASS 2
Voltage, electrical, arc flash
30.48 cm
17000/25500
Leather, Rubber
12 in.
1
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit