Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ ArcGuard™ Class 2 Rubber Voltage Gloves Premium Kit
SDP

Click to view available options
Size:
8
9
10
11
12

Premium kit includes a pair of electrical insulating rubber gloves, fire-resistant knit gloves, leather glove protectors, and a glove bag. Class 2 insulating gloves will protect workers from up to 17,000 V.

  • Electrical insulating gloves protect against electrical shock and other arc flash hazards
  • Wear with included leather glove protectors for additional resistance against mechanical hazards
  • FR knit gloves stretch for dexterity and can be worn for another layer of protection
  • Compliant with ANSI/ASTM D120 and NFPA 70E standards
  • Class 2 gloves offer protection from up to 17,000 V
  • Gloves should be used within one year of marked test date
  • When not in use, lay flat in the included protective bag
  • Store away from direct sunlight, high heat, and humidity
Certifications/Compliance ANSI/ASTM D120, Class 2, Type I, NFPA 70E-2015
Applications Industrial
Color Black
Material Rubber, Leather
Product Type Electrical Insulating Gloves Kit
Size 12
For Use With (Application) Electrical Utility Work, Shock Protection
Glove Cuff Style Rolled
Includes Electrical Insulating Rubber Gloves, Fire-Resistant Knit Gloves, Leather Glove Protectors, and a Glove Bag
Series KITGC2BAG
Voltage 17,000 V
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit