Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
National Safety Apparel™ ArcGuard™ Class 2 Rubber Voltage Gloves Premium Kit

Premium kit includes a pair of electrical insulating rubber gloves, fire-resistant knit gloves, leather glove protectors, and a glove bag. Class 2 insulating gloves will protect workers from up to 17,000 V.
$395.25
Specifications
Certifications/Compliance | ANSI/ASTM D120, Class 2, Type I, NFPA 70E-2015 |
---|---|
Color | Black |
Glove Cuff Style | Rolled |
Includes | Electrical Insulating Rubber Gloves, Fire-Resistant Knit Gloves, Leather Glove Protectors, and a Glove Bag |
Material | Rubber, Leather |
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
17-355-279
![]() |
National Safety Apparel™
KITGC2B08AG |
8 |
Each for $395.25
|
|
17-355-281
![]() |
National Safety Apparel™
KITGC2B09AG |
9 |
Each for $395.25
|
|
17-355-283
![]() |
National Safety Apparel™
KITGC2B10AG |
10 |
Each for $395.25
|
|
17-355-285
![]() |
National Safety Apparel™
KITGC2B11AG |
11 |
Each for $395.25
|
|
17-355-287
![]() |
National Safety Apparel™
KITGC2B12AG |
12 |
Each for $395.25
|
|
Description
- Electrical insulating gloves protect against electrical shock and other arc flash hazards
- Wear with included leather glove protectors for additional resistance against mechanical hazards
- FR knit gloves stretch for dexterity and can be worn for another layer of protection
- Compliant with ANSI/ASTM D120 and NFPA 70E standards
- Class 2 gloves offer protection from up to 17,000 V
- Gloves should be used within one year of marked test date
- When not in use, lay flat in the included protective bag
- Store away from direct sunlight, high heat, and humidity
Specifications
ANSI/ASTM D120, Class 2, Type I, NFPA 70E-2015 | |
Rolled | |
Rubber, Leather | |
KITGC2BAG |
Black | |
Electrical Insulating Rubber Gloves, Fire-Resistant Knit Gloves, Leather Glove Protectors, and a Glove Bag | |
Electrical Insulating Gloves Kit | |
17,000 V |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title