Learn More
National Safety Apparel™ ArcGuard™ Class 2 Rubber Voltage Gloves Premium Kit
Premium kit includes a pair of electrical insulating rubber gloves, fire-resistant knit gloves, leather glove protectors, and a glove bag. Class 2 insulating gloves will protect workers from up to 17,000 V.
Manufacturer:  National Safety Apparel™ KITGC208AG
 View more versions of this product
Catalog No. 17-001-2001
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description
Description
  • Electrical insulating gloves protect against electrical shock and other arc flash hazards
  • Wear with included leather glove protectors for additional resistance against mechanical hazards
  • FR knit gloves stretch for dexterity and can be worn for another layer of protection
  • Compliant with ANSI/ASTM D120 and NFPA 70E standards
  • Class 2 gloves offer protection from up to 17,000 V
  • Gloves should be used within one year of marked test date
  • When not in use, lay flat in the included protective bag
  • Store away from direct sunlight, high heat, and humidity
Compliance
ANSI/ASTM D120, Class 2, Type I; NFPA 70E-2015
Specifications
Specifications
Electrical Insulating Gloves Kit
Electrical Utility Work, Shock Protection
Electrical Insulating Rubber Gloves, Fire-Resistant Knit Gloves, Leather Glove Protectors, and a Glove Bag
KITGC2BAG
17,000 V
Industrial
Black
Rolled
Rubber, Leather
8
ANSI/ASTM D120, Class 2, Type I, NFPA 70E-2015