Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ ArcGuard™ Class 2 Rubber Voltage Gloves Premium Kit

Premium kit includes a pair of electrical insulating rubber gloves, fire-resistant knit gloves, leather glove protectors, and a glove bag. Class 2 insulating gloves will protect workers from up to 17,000 V.

Specifications

Certifications/Compliance ANSI/ASTM D120, Class 2, Type I, NFPA 70E-2015
Color Black
Glove Cuff Style Rolled
Includes Electrical Insulating Rubber Gloves, Fire-Resistant Knit Gloves, Leather Glove Protectors, and a Glove Bag
Material Rubber, Leather
View More Specs

Products 5
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
17-001-2001
View Documents
National Safety Apparel™
KITGC208AG
8 This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
17-001-2002
View Documents
National Safety Apparel™
KITGC209AG
9 This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
17-001-2003
View Documents
National Safety Apparel™
KITGC210AG
10 This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
17-001-2004
View Documents
National Safety Apparel™
KITGC211AG
11 This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
17-001-2005
View Documents
National Safety Apparel™
KITGC212AG
12 This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description

Description

  • Electrical insulating gloves protect against electrical shock and other arc flash hazards
  • Wear with included leather glove protectors for additional resistance against mechanical hazards
  • FR knit gloves stretch for dexterity and can be worn for another layer of protection
  • Compliant with ANSI/ASTM D120 and NFPA 70E standards
  • Class 2 gloves offer protection from up to 17,000 V
  • Gloves should be used within one year of marked test date
  • When not in use, lay flat in the included protective bag
  • Store away from direct sunlight, high heat, and humidity

Compliance

ANSI/ASTM D120, Class 2, Type I; NFPA 70E-2015

Specifications

Specifications

ANSI/ASTM D120, Class 2, Type I, NFPA 70E-2015
Rolled
Rubber, Leather
KITGC2BAG
Black
Electrical Insulating Rubber Gloves, Fire-Resistant Knit Gloves, Leather Glove Protectors, and a Glove Bag
Electrical Insulating Gloves Kit
17,000 V
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit