Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
National Safety Apparel™ ArcGuard™ Class 2 Rubber Voltage Gloves
Click to view available options
Color:
Black
Red
Yellow
Size:
8
9
10
11
12
Description
- Class 2 rubber electrical insulating gloves
- Compliant with ANSI/ASTM D120 and NFPA 70E-2015 standards for use around electrical and arc flash hazards
- Gloves should be paired with 10 in. leather glove protectors for additional protection against mechanical hazards
- Must be used within 12 months of the stamped test date
Specifications
Certifications/Compliance | ANSI/ASTM D120, Class 2, Type I, NFPA 70E-2015 |
Applications | Industrial |
Color | Yellow |
Material | Rubber |
Size | 9 |
For Use With (Application) | Electrical Utility Work |
Glove Cuff Style | Rolled |
Series | GC2Y |
Length (English) | 14 in. |
Length (Metric) | 35.5 cm |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction