Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pyramex™ Class 3 Weatherproof Parkas

Pyramex™ Class 3 Weatherproof Parkas with ANSI/ISEA 107-2015 safety compliance
$105.00
Specifications
Material | Polyester |
---|---|
Closure Type | Zipper |
Certifications/Compliance | ANSI/ISEA 107-2015, Type R - Class 3 |
Coating | Polyurethane |
Cuff Style | Elastic |
Catalog Number | Mfr. No. | Color | Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Size | Price | Quantity & Availability | ||||
19-555-322
![]() |
Pyramex™
RP3110M |
Hi-vis Lime | Medium |
Each for $105.00
|
|
19-555-321
![]() |
Pyramex™
RP3110L |
Hi-vis Lime | Large |
Each for $105.00
|
|
19-555-327
![]() |
Pyramex™
RP3110XL |
Hi-vis Lime | X-Large |
Each for $105.00
|
|
19-555-323
![]() |
Pyramex™
RP3110X2 |
Hi-vis Lime | 2X-Large |
Each for $105.00
|
|
19-555-324
![]() |
Pyramex™
RP3110X3 |
Hi-vis Lime | 3X-Large |
Each for $105.00
|
|
19-555-325
![]() |
Pyramex™
RP3110X4 |
Hi-vis Lime | 4X-Large |
Each for $105.00
|
|
19-555-326
![]() |
Pyramex™
RP3110X5 |
Hi-vis Lime | 5X-Large |
Each for $105.00
|
|
19-555-329
![]() |
Pyramex™
RP3120M |
Hi-vis Orange | Medium |
Each for $105.00
|
|
19-555-328
![]() |
Pyramex™
RP3120L |
Hi-vis Orange | Large |
Each for $105.00
|
|
19-555-334
![]() |
Pyramex™
RP3120XL |
Hi-vis Orange | X-Large |
Each for $105.00
|
|
19-555-330
![]() |
Pyramex™
RP3120X2 |
Hi-vis Orange | 2X-Large |
Each for $105.00
|
|
19-555-331
![]() |
Pyramex™
RP3120X3 |
Hi-vis Orange | 3X-Large |
Each for $105.00
|
|
19-555-332
![]() |
Pyramex™
RP3120X4 |
Hi-vis Orange | 4X-Large |
Each for $105.00
|
|
19-555-333
![]() |
Pyramex™
RP3120X5 |
Hi-vis Orange | 5X-Large |
Each for $105.00
|
|
Description
- Weatherproof
- Hi-Vis polyester shell with zip out polar fleece liner
- Elastic band at the wrists
- 2 in. silver reflective material
- Concealed detachable hood with drawstring
- Zipper front closure
- Interior access zipper for print customization
- Cell phone/radio chest pocket with clear panel and 2 mic tabs
- Drawstring around bottom
Specifications
Polyester | |
ANSI/ISEA 107-2015, Type R - Class 3 | |
Elastic | |
Polar Fleece | |
Weatherproof |
Zipper | |
Polyurethane | |
Concealed, Detachable | |
8 | |
China Brand Silver |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title