Learn More
United Scientific™ Class A Volumetric Pipette, Individually Certified
SDPSCI_ED
Glass Pipettes featuring clear markings and calibrated 'To Deliver'.
Supplier:  United Scientific™ PT7100G
 View more versions of this product
Catalog No. S01025
Explore available promotions
Add to Cart
This item is not returnable. View return policy
Description
Description
  • Made from borosilicate glass in compliance with ASTM specification E969
  • Color-coded
  • Individually serialized and certified
  • Calibrated to deliver
  • Printed with durable amber markings
Specifications
Specifications
Red Color Code
Non-sterile
0.05 mL
50 mL
Reusable
Borosilicate Glass
Yes
Clear
Volumetric Pipet
Pipet Filler
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
Warning: Cancer and reproductive harm