Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medique Class B 50-Person First-Aid Kit

Provides first aid for up to 50 people.

$152.00 - $186.70

Specifications

Product Type Class B First Aid Kit
Sufficient For 50 Persons
View More Specs

Products 2
Catalog Number Material Price Quantity & Availability  
Catalog Number Material Price Quantity & Availability  
19-090-669
View Documents
Plastic
Each for $152.00
Case of 10 EA for $1,362.00
Add to Cart
 
19-090-670
View Documents
Metal
Each for $186.70
Case of 6 EA for $1,001.00
Add to Cart
 
Description

Description

  • For small office setting
  • Wall mounted for ease of use
Specifications

Specifications

Class B First Aid Kit
50 Persons
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit