Learn More
Eisco™ Class B Glass Graduated Cylinders, ASTM
SCI_ED
Class B glass graduated cylinders are ASTM compliant and feature a hexagonal base, printed graduations, and spout.
Manufacturer:  Eisco™ CH0341MBL
 View more versions of this product
Catalog No. S27919
Add to cart
Description
Description
  • Made from high quality borosilicate 3.3 glass
  • Hexagonal base provides additional stability
  • Printed graduations are chemical resistant
  • Conforms to ASTM E1272 standard
  • Spout for easy, mess free pouring
  • Autoclavable
  • Class B
Specifications
Specifications
Blue
Hexagonal
B
Graduated Cylinder
250 mL
Autoclavable
30.5 cm
ASTM E1272
12 in.
Borosilicate Glass
2 mL