Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clathrin Heavy Chain 1/CHC17 Mouse anti-Human, Mouse, Rat, Porcine, Avian, Bovine, Canine, Monkey, Xenopus, Clone: CHC/1432, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen Clathrin Heavy Chain 1/CHC17
Clone CHC/1432
Concentration 0.2 mg/ml
Dilution Immunocytochemistry/Immunofluorescence 1-2 ug/ml
Applications Immunofluorescence
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122612
View Documents
Novus Biologicals
NBP30756920UG
20 μg
Each for $138.37
Add to cart
 
NB122611
View Documents
Novus Biologicals
NBP30756910UG
100 μg
Each for $352.89
Add to cart
 
Description

Description

Clathrin Heavy Chain 1/CHC17 Monoclonal antibody specifically detects Clathrin Heavy Chain 1/CHC17 in Human, Mouse, Rat, Porcine, Avian, Bovine, Canine, Monkey, Xenopus samples. It is validated for Immunofluorescence
Specifications

Specifications

Clathrin Heavy Chain 1/CHC17
0.2 mg/ml
Immunofluorescence
Purified
RUO
Human, Mouse, Rat, Porcine, Avian, Bovine, Canine, Monkey, Xenopus
CHC, CHC17, clathrin heavy chain 1, Clathrin heavy chain on chromosome 17, clathrin, heavy chain, clathrin, heavy chain (Hc), clathrin, heavy polypeptide (Hc), clathrin, heavy polypeptide-like 2, CLH17, CLH-17, CLTCL2, Hc, KIAA0034
Recombinant full-length human Clathrin Heavy Chain 1/CHC17 protein
Monoclonal
Protein A or G purified
CHC/1432
Immunocytochemistry/Immunofluorescence 1-2 ug/ml
Unconjugated
Mouse
Membrane Trafficking and Chaperones, Membrane Vesicle Markers
10 mM PBS with 0.05% BSA
1213
IgG1 Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit