Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clathrin Heavy Chain 1/CHC17 Mouse anti-Human, Mouse, Rat, Porcine, Bovine, PE-Cyanine7, Clone: BF-06, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP145050PECY7

Catalog No. N145050PEC7

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Clathrin Heavy Chain 1/CHC17 Monoclonal antibody specifically detects Clathrin Heavy Chain 1/CHC17 in Human, Mouse, Rat, Porcine, Bovine samples. It is validated for Flow Cytometry.
Specifications

Specifications

Clathrin Heavy Chain 1/CHC17
BF-06
Flow Cytometry
Conjugated
CLTC
The antibody BF-06 recognizes clathrin heavy chain, an ubiquitously expressed 180 kDa protein involved in receptor-mediated endocytosis.
Ion exchange chromatography
Membrane Trafficking and Chaperones, Membrane Vesicle Markers
Primary
1213.0
Bovine, Human, Mouse, Porcine, Rat
Flow Cytometry
PE-Cyanine7
PBS with 0.05% Sodium Azide
CHC, CHC17, clathrin heavy chain 1, Clathrin heavy chain on chromosome 17, clathrin, heavy chain, clathrin, heavy chain (Hc), clathrin, heavy polypeptide (Hc), clathrin, heavy polypeptide-like 2, CLH17, CLH-17, CLTCL2, Hc, KIAA0034
Mouse
IgM
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
The antibody BF-06 recognizes clathrin heavy chain, an ubiquitously expressed 180 kDa protein involved in receptor-mediated endocytosis.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit