Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clathrin interactor 1 Mouse anti-Human, Alexa Fluor™ 647, Clone: OTI8C1, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271491AF647

Catalog No. N271491A647


Add to cart

Description

Description

Clathrin interactor 1 Monoclonal antibody specifically detects Clathrin interactor 1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

Clathrin interactor 1
OTI8C1
Western Blot
Affinity Purified, Conjugated
CLINT1
Recombinant protein fragment corresponding to amino acids 1-290 of human Clathrin interactor 1 (NP_055481) produced in E.coli.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Human
Western Blot
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
clathrin interacting protein localized in the trans-Golgi region, clathrin interactor 1, Clathrin-interacting protein localized in the trans-Golgi region, CLINT, ENTHClint, enthoprotin, EPN4Enthoprotin, EPNRepsin 4, Epsin-4, EpsinR, Epsin-related protein, FLJ46753, KIAA0171epsinR
Mouse
IgG1
0.1 mL
Primary
9685.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit