Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clathrin light chain Mouse anti-Human, Allophycocyanin/Cy7, Clone: SPM174, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP254435APCCY7

Catalog No. N254435APC7


Add to cart

Description

Description

Clathrin light chain Monoclonal antibody specifically detects Clathrin light chain in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

Clathrin light chain
SPM174
Flow Cytometry
Conjugated, Purified
CLTA
Purified recombinant N-terminal fragment of human Clathrin Light Chain
Protein A or G purified
Membrane Trafficking and Chaperones, Membrane Vesicle Markers
Primary
1211.0
Flow Cytometry
APC-Cyanine7
PBS with 0.05% Sodium Azide
clathrin light chain A, clathrin, light chain A, LCA, light polypeptide (Lca)
Mouse
IgG2b Kappa
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit