Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clathrin light chain Mouse anti-Human, Clone: CLC/1421, Novus Biologicals™

Mouse Monoclonal Antibody

$152.22 - $378.35

Specifications

Antigen Clathrin light chain
Clone CLC/1421
Host Species Mouse
Isotype IgG2b Kappa
Gene Alias clathrin light chain A, clathrin, light chain A, LCA, light polypeptide (Lca)
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB25323120 View Documents Novus Biologicals
NBP25323120UG
20 ug Each for $152.22
Add to cart
 
NB253231100 View Documents Novus Biologicals
NBP253231100UG
100 ug Each for $378.35
Add to cart
 
NB253231200 View Documents Novus Biologicals
NBP253231200UG
200 ug This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Clathrin light chain Monoclonal antibody specifically detects Clathrin light chain in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, SDS-Page.
Specifications

Specifications

Clathrin light chain
Mouse
clathrin light chain A, clathrin, light chain A, LCA, light polypeptide (Lca)
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), SDS-Page, Western Blot
Unconjugated
Store at 4C.
Monoclonal
0.2 mg/mL
CLC/1421
IgG2b Kappa
Human
RUO
Purified
Primary
10mM PBS with 0.05% BSA with 0.05% Sodium Azide

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit