Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Clathrin Light Chain Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5100442
Description
Antibody detects endogenous levels of total CLTB.
Clathrin is the major protein of the polyhedral coat of coated pits and vesicles. Two different adapter protein complexes link the clathrin lattice either to the plasma membrane or to the trans-Golgi network.
Specifications
Clathrin Light Chain | |
Polyclonal | |
Unconjugated | |
CLTB | |
2310046E19Rik; AV026556; CHC17; clathrin light chain A; clathrin light chain A S homeolog; clathrin light chain A S homeolog; clathrin, light chain A S homeolog; clathrin light chain B; clathrin, light chain (Lca); clathrin, light chain (Lcb); clathrin, light chain A; clathrin, light chain A2; clathrin, light chain B; clathrin, light polypeptide (Lca); clathrin, light polypeptide (Lcb); clathrin, light polypeptide A b; clathrin, light polypeptide B (light chain B); clathryn light chain (LCA2); CLH17; clta; clta.S; CLTA2; clta-a; cltab; clta-b; CLTB; CLTC; CLTLB; CTLA; CTLB; KIAA0034; lca; LCA1; LCA2; LCA3; Lcb; unnamed protein product; wu:fb61d08; wu:fj48d06; XELAEV_18001213mg; zgc:73358 | |
Rabbit | |
Affinity chromatography | |
RUO | |
116561, 1211, 1212, 12757, 74325, 83800 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O08585, P08081, P08082, P09496, P09497, Q6IRU5 | |
CLTA, CLTB | |
A synthesized peptide derived from human CLTB(Accession P09497), corresponding to amino acid residues S61-R111. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction