Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ClathrinHeavyChain2/CHC22/CLTCL1 Rabbit anti-Human, Clone: JA53-02, Novus Biologicals™
Rabbit Monoclonal Antibody
Manufacturer: Novus Biologicals NBP266781
Description
ClathrinHeavyChain2/CHC22/CLTCL1 Monoclonal antibody specifically detects ClathrinHeavyChain2/CHC22/CLTCL1 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
ClathrinHeavyChain2/CHC22/CLTCL1 | |
JA53-02 | |
Unconjugated | |
Purified | |
CHC22, clathrin heavy chain 2, clathrin heavy chain on chromosome 22, clathrin, heavy chain-like 1, Clathrin, heavy polypeptide D, CLTCL, heavy polypeptide-like 1 | |
CLTCL1 | |
Recombinant Protein | |
Monoclonal | |
Protein A purified | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot,Immunohistochemistry,Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Western Blot 1:100-1:2000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin | |
TBS (pH7.4), 1%BSA, 40%Glycerol with 0.05% Sodium Azide | |
8218 | |
Rabbit | |
IgG | |
Primary | |
100μL | |
Human,Mouse,Rat |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title