Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Claudin 1, Rabbit Polyclonal Antibody

Specifically detect Claudin 1 in human, monkey, mouse, dog and rat samples with Thermo Scientific™ Claudin 1, Rabbit Polyclonal Antibody.

Manufacturer:  Thermo Scientific™ RB9209P0

 View more versions of this product

Catalog No. RB9209P0

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Specifically detect Claudin 1 in human, monkey, mouse, dog and rat samples with Thermo Scientific™ Claudin 1, Rabbit Polyclonal Antibody.

Claudin 1-8 proteins are a family of transmembrane proteins associated with tight junctions. Tight junctions are specialized regions of cell to cell contact; made up of network of strands to act as a molecular gasket for preventing the leakage of ions, water etc. between cells. They are abundant in luminal epithelial sheets where they maintain epithelial cell polarity. Different tissues exhibit different Claudin composition.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Monkey, Mouse, Dog and Rat. Others not tested.

Epitope: C-terminal

Immunogen: A synthetic peptide derived from the C-terminus of human Claudin 1

Molecular Weight: 22 kDa

Positive Control: T47D cells, Skin or Breast CA

Cellular Localization: Membrane on skin and cytoplasmic and nuclear on breast carcinoma

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
Specifications

Specifications

Claudin 1
Polyclonal
Unconjugated
Concentrated
A synthetic peptide derived from the C-terminus of human Claudin-1
100μL
Cell Adhesion
Primary
Canine, Human, Murine, Non-human Primate, Rat
Immunohistochemistry (Paraffin), Western Blot
200µg/mL
Purified Ab with BSA and Azide
Rabbit
IgG
IVD
Primary
Polyclonal
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit