Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Claudin-16 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP15910520UL
Description
Claudin-16 Polyclonal specifically detects Claudin-16 in Human, Mouse samples. It is validated for Western Blot.Specifications
Claudin-16 | |
Polyclonal | |
Western Blot 1:100-1:2000 | |
Q9Y5I7 | |
CLDN16 | |
Synthetic peptides corresponding to CLDN16 (claudin 16) The peptide sequence was selected from the C terminal of CLDN16. Peptide sequence FLAGAVLTCCLYLFKDVGPERNYPYSLRKAYSAAGVSMAKSYSAPRTETA. | |
20 μL | |
Primary | |
Human | |
Purified |
Western Blot | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
claudin 16, claudin-16, HOMG3paracellin-1, Paracellin-1, PCLN-1, PCLN1hypomagnesemia 3, with hypercalciuria and nephrocalcinosis | |
Rabbit | |
Protein A purified | |
RUO | |
10686 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title