Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Claudin-3 Antibody (OTI1E7), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270445AF700
Description
Claudin-3 Monoclonal specifically detects Claudin-3 in Human, Mouse, Rat, Zebrafish samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Claudin-3 | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CLDN3 | |
Full length recombinant protein of human Claudin-3 (NP_001297) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Zebrafish | |
IgG2b |
Western Blot, Immunohistochemistry | |
OTI1E7 | |
Western Blot, Immunohistochemistry | |
C7orf1, claudin 3, claudin-3, Clostridium perfringens enterotoxin receptor 2, CPE-R 2, CPE-R2, CPETR2CPE-receptor 2, HRVP1, Rat ventral prostate.1 protein homolog, RVP1, ventral prostate.1 protein homolog, ventral prostate.1-like protein | |
Mouse | |
Affinity Purified | |
Cell Biology, Cytoskeleton Markers, Extracellular Matrix | |
1365.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction