Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Claudin-3, Mouse anti-Human, Clone: 385013, R&D Systems™

Mouse Monoclonal Antibody

$119.60 - $324.50

Specifications

Antigen Claudin-3
Clone 385013
Immunogen HEK293 human embryonic kidney cell line transfected with human Claudin-3, Met1-Val220, Accession # O15551
Host Species Mouse
Primary or Secondary Primary
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB46201100
View Documents
R&D Systems
MAB46201100
100 ug
Each for $324.50
Add to cart
 
MAB46201SP
View Documents
R&D Systems
MAB46201SP
25 ug
Each for $119.60
Add to cart
 
Description

Description

Claudin-3 Monoclonal antibody specifically detects Claudin-3 in Human Samples. It is validated for Flow Cytometry, CyTOF.
Specifications

Specifications

Claudin-3
HEK293 human embryonic kidney cell line transfected with human Claudin-3, Met1-Val220, Accession # O15551
Primary
Human
Unconjugated
1365
Purified
Flow Cytometry, CyTOF
RUO
385013
Mouse
Monoclonal
IgG2a
C7orf1, claudin 3, claudin-3, Claudin3, CLDN3, Clostridium perfringens enterotoxin receptor 2, CPE-R 2, CPE-R2, CPETR2, CPETR2CPE-receptor 2, HRVP1, Rat ventral prostate.1 protein homolog, RVP1, ventral prostate.1 protein homolog, ventral prostate.1-like protein
Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit