Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Claudin 3, Rabbit Polyclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ Claudin 3, Rabbit Polyclonal Antibody.

Manufacturer:  Thermo Scientific™ RB9251P1

 View more versions of this product

Catalog No. RB9251P1

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Obtain accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ Claudin 3, Rabbit Polyclonal Antibody, one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available.

Claudin 1-8 proteins are a family of transmembrane proteins associated with tight junctions. Tight junctions are specialized regions of cell to cell contact; made up of network of strands to act as a molecular gasket for preventing the leakage of ions, water etc. between cells. They are abundant in luminal epithelial sheets where they maintain epithelial cell polarity. Different tissues exhibit different Claudin composition.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse and Rat. Others not tested.

Epitope: C-terminal

Immunogen: A synthetic peptide derived from the C-terminus of mouse Claudin 3

Molecular Weight: 22-24 kDa

Positive Control: Small intestine

Cellular Localization: Membrane and cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

Claudin 3
Polyclonal
Unconjugated
Rabbit
IgG
IVD
Primary
Polyclonal
Immunohistochemistry (Paraffin)
200µg/mL
Concentrated
A synthetic peptide derived from the C-terminus of mouse Claudin-3
500μL
Cell Adhesion
Primary
Human, Murine
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit