Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Claudin-5 Mouse anti-Human, Alexa Fluor™ 405, Clone: OTI1G4, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271329AF405

Catalog No. N271329A405


Add to cart

Description

Description

Claudin-5 Monoclonal antibody specifically detects Claudin-5 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

Claudin-5
OTI1G4
Flow Cytometry, Immunocytochemistry/Immunofluorescence
Affinity Purified, Conjugated
CLDN5
Full length recombinant protein of human Claudin-5 (NP_003268) produced in HEK293T cells.
Immunogen affinity purified
Cellular Markers
Primary
7122.0
Flow Cytometry, Immunocytochemistry, Immunofluorescence
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
AWALTransmembrane protein deleted in VCFS, BEC1, claudin 5, claudin-5, CPETRL1, TMVCFTMDVCF, transmembrane protein deleted in velocardiofacial syndrome
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit