Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Claudin 5 Mouse anti-Human, Unconjugated, Clone: 2B12, MilliporeSigma™

Mouse Monoclonal Antibody

$147.00 - $372.00

Specifications

Antigen Claudin 5
Applications ELISA, Flow Cytometry, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Mouse
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABT15281MI
View Documents
MilliporeSigma
MABT1528100UL
100 μL
Each for $372.00
Add to cart
 
MABT15282MI
View Documents
MilliporeSigma
MABT152825UL
25 μL
Each for $147.00
Add to cart
 
Description

Description

Claudin 5 Monoclonal antibody specifically detects Claudin 5 in Human samples. It is validated for ELISA, Flow Cytometry, Inhibits Activity/Function, Western Blotting.
Specifications

Specifications

Claudin 5
Unconjugated
Mouse
Transmembrane protein deleted in VCFS;TMDVCF
An artificial membrane protein consisting of the extracellular region of human Claudin 5.
Primary
Stable for 1 year at -20°C from date of receipt.
ELISA, Flow Cytometry, Western Blot
Purified
Human
CLDN5, AWAL, TMVCF
Monoclonal
Protein G Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit