Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Claudin-7 Mouse anti-Human, Clone: 6-G3, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP266784
Description
Claudin-7 Monoclonal antibody specifically detects Claudin-7 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
Claudin-7 | |
6-G3 | |
Unconjugated | |
Purified | |
CEPTRL2, claudin 7, claudin-1, claudin-7, CLDN-7, clostridium perfringens enterotoxin receptor-like 2, CPETRL2 | |
CLDN7 | |
Peptide | |
Monoclonal | |
Protein A purified | |
Breast Cancer, Cancer | |
Human,Mouse,Rat |
Immunocytochemistry,Immunofluorescence,Western Blot | |
2 mg/mL | |
Western Blot 1:500-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:200 | |
TBS (pH7.4), 1%BSA, 40% Glycerol with 0.05% Sodium Azide | |
1366 | |
Mouse | |
IgG1 | |
Primary | |
100μL | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title