Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Claudin 7 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Manufacturer: Proteintech Group Inc 101181AP150UL

Description
This gene encodes a member of the claudin family. Claudins are integral membrane proteins and components of tight junction strands. Tight junction strands serve as a physical barrier to prevent solutes and water from passing freely through the paracellular space between epithelial or endothelial cell sheets, and also play critical roles in maintaining cell polarity and signal transductions. Differential expression of this gene has been observed in different types of maligcies, including breast cancer, ovarian cancer, hepatocellular carcinomas, urinary tumors, prostate cancer, lung cancer, head and neck cancers, thyroid carcinomas, etc. Alternatively spliced transcript variants encoding different isoforms have been found.Specifications
Claudin 7 | |
Polyclonal | |
Unconjugated | |
CLDN7 | |
CEPTRL2, Claudin 7, CLDN 7, CLDN7, CPETRL2, Hs.84359 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1366 | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin) | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O95471 | |
CLDN7 | |
Claudin 7 Fusion Protein Ag0159 | |
150 μL | |
Primary | |
Human | |
Liquid |
Safety and Handling
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title