Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Claudin 8 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA124415
Change view
Click to view available options
Quantity:
100 μg
Catalog No. Quantity
PIPA124415 100 μg
1 options

Catalog No. PIPA124415

Supplier: Thermo Scientific PA124415

Rabbit Polyclonal Antibody

PA1-24415 detects Claudin 8 in Human samples. PA1-24415 has been successfully used in Western blot and immunohistochemistry(paraffin) procedures. The PA1-24415 immunogen is a synthetic peptide region of human CLDN8. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

CLDN8, clustered with CLDN17 at human chromosome 21q22.11, is a four-transmembrane protein with WWCC motif, defined by W-X(17-22)-W-X(2)-C-X(8-10)-C.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Claudin 8
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Gene CLDN8
Gene Accession No. P56748, Q9Z260
Gene Symbols CLDN8
Host Species Rabbit
Immunogen Synthetic peptide region of human CLDN8.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54420, 9073
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.