Learn More
Claudin 9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 161961AP
Catalog No. 50-173-0782
Add to cart
Description
Description
This antibody is specifically against claudin 9.
CLDN9 is a member of claudin family. It is a four-transmembrane protein with WWCC motif, defined by W-X(17-22)-W-X(2)-C-X(8-10)-C.
Specifications
Specifications
Claudin 9
0.15 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O95484, Q9Z0S7
claudin 9, CLDN9
Rabbit
IgG
150 μL
-20°C
Primary
287099, 56863, 9080
Immunoprecipitation, Western Blot
Unconjugated
CLDN9
Liquid
Cldn9
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat