Learn More
BD Clay Adams 100mL Capacity 4-Place Horizontal Rotor for Dynac and Dynac III Centrifuges
Manufacturer:  BD 420110
Catalog No. 05-100-30
Add to cart
Description
Description
4 × 100mL capacity. Operates at speeds to 2450rpm, creating forces of 1150×G in Dynac centrifuge and 1215×G in Dynac III models. For use with Dynac, Dynac II, and Dynac III Centrifuges.
Order Info
Accepts 100mL tubes when equipped with 05-100-35 bucket (sold separately).
Specifications
Specifications
Aluminum
4 x 100 mL
2,385/2,450 rpm
Trunnion rings
Horizontal Rotor
1,150/1,215 x g
Dynac™, Dynac™ II and Dynac™ III Centrifuge