Learn More
BD Clay Adams 4-Place Fixed-Angle Rotor for Dynac and Dynac III Centrifuges
Manufacturer:  BD 420112
Catalog No. 05-100-32
Add to cart
Description
Description
4 × 50mL capacity. This 52° fixed-angle rotor accommodates loads up to 200mL when equipped with 05-100-36 bucket (sold separately). For use with Dynac, Dynac II, and Dynac III Centrifuges.
Specifications
Specifications
Aluminum
4 x 50 mL
2,860 rpm
52°
Fixed Angle Rotor
1,508 x g
Dynac™ and Dynac™ III Centrifuge