Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLC-5 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA564891

Catalog No. PIPA564891


Add to cart

Description

Description

Immunogen sequence: MAMWQGAMDNR GFQQGSFSSF QNSSSDEDLM DIPATAMDFS MRDDVPPLDR EVG Highest antigen sequence identity to the following orthologs - mouse 91%, rat 91%.

Proton-coupled chloride transporter. Functions as antiport system and exchanges chloride ions against protons. Important for normal acidification of the endosome lumen. May play an important role in renal tubular function.
Specifications

Specifications

CLC-5
Polyclonal
Unconjugated
CLCN5
Liquid
CLCN5
Recombit Human CLCN5
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Immunocytochemistry, Immunofluorescence, Western Blot
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
P51795
CLC5; CLCK2; DENTS; NPHL1; NPHL2; XLRH; XRN; hCIC-K2; hClC-K2; H(+)/Cl(-) exchange transporter 5; chloride channel protein 5; chloride transporter ClC-5; clC-5
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
1184
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only