Learn More
Invitrogen™ CLCNKA Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA592064
Description
Immunogen sequence: QSCQPSFYDG TIIVKKLPYL PRILGRNIGS HHVRVEHFMN HSITTLAKDT PLEEVVKVVT STDVTEYPLV ESTESQILVG IVQRAQLVQA LQAEPPSRAP GHQQCLQDIL ARGCPTEPVT LTLFSETTLH QAQNLFKLLN LQSLFVTSRG RAVGCVSWVE MKKAISNLTN PPAPK; Positive Samples: LO2, HT-29, A-549; Cellular Location: Membrane, Multi-pass membrane protein.
CLC-KA is a kidney-specific chloride channel that mediates transepithelial chloride transport in the thin ascending limb of the Henle loop in the inner medulla. CLC-KA plays a crucial role in urine concentration. The gene encoding human CLC-KA maps to chromosome 1p36. Mutations in this gene may be associated with nephrogenic diabetes insipidus in those cases where mutations in the vasopressin V2 receptor and the AQP2 water channel are lacking. CLC-KB mediates basolateral chloride ion efflux in the thick ascending limb and in more distal nephron segments. The gene encoding human CLC-KB maps to chromosome 1p36. Mutations in this gene cause type III Barter's syndrome which is characterized by renal salt-wasting and low blood pressure.
Specifications
CLCNKA | |
Polyclonal | |
Unconjugated | |
CLCNKA | |
C75963; chloride channel K1; chloride channel Ka; Chloride channel protein ClC-Ka; chloride channel, kidney, A; chloride channel, voltage-sensitive Ka; chloride voltage-gated channel Ka; CLCK1; clC-K1; CLCKA; CLC-KA; Clcnk1; Clcnka; hClC-Ka; putative basolateral cTAL chloride channel ClC-Ka | |
Rabbit | |
Affinity Chromatography | |
RUO | |
1187, 12733 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.18 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
P51800, Q9WUB7 | |
CLCNKA | |
Recombinant fusion protein containing a sequence corresponding to amino acids 470-644 of human CLCNKA (NP_0012440681). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.