Learn More
Peptides InternationalSupplier Diversity Partner CLE25 Peptide (0.5 mg vial), MF= C58H94N20O19 DFS Item
Manufacturer:  Peptides InternationalSupplier Diversity Partner PBP4511V 1EA
CLAVATA3/ESR-Related 25 Peptide; Arg-Lys-Val-Hyp-Asn-Gly-Hyp-Asp-Pro-Ile-His-Asn, Application: Root-to-Shoot Transmitter of Water-Deficiency Signals, origin/species: Arabidopsis thaliana, MW: 1375.5
Catalog No. 50-195-2416