Learn More
Clean Clay Loam, MilliporeSigma™ Supelco™
Clean clay loam may be used as a reference material in the recovery of nitro-organic explosives in soil samples using gas chromatography coupled with electron capture detector (GC-ECD).
$148.10 - $288.00
Specifications
CAS 14464-46-1
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
11-100-1668
View Documents
MilliporeSigma Supelco
CLNSOIL2100G
100 g
Each for $148.10
Add to cart
11-100-1665
View Documents
MilliporeSigma Supelco
CLNSOIL2250G
250 g
Each for $288.00
Add to cart
Description
Description
Featured Industries: Environmental
Specifications
Specifications
14464-46-1