Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

Stone Medical Corporation Clean Collect™ Diversion Blood Culture Collection Kit for BacTAlert Blood Culture Bottles

Prevent blood culture contamination with a diversion system peripheral blood culture kit.

$206.00 - $254.93

Specifications

For Use With (Application) BacTAlert Blood Culture Bottles, BacTAlert Blood Culture Bottles
Quantity 40/Cs, 40/Cs
Sample Type Blood, Blood
Storage Requirements 5 to 25°C, 5 to 25°C
View More Specs

Products 2
Catalog Number Mfr. No. Includes Price Quantity & Availability  
Catalog Number Mfr. No. Includes Price Quantity & Availability  
23-200-116
View Documents
Stone Medical Corporation
CC225G
Greiner Butterfly Needle Case of 40 for $206.00
N/A
23-200-121
View Documents
Stone Medical Corporation
CC225D
BD Slide Butterfly Needle Case of 40 for $254.93
N/A
Description

Description

  • Reduces contamination
  • Improves patient outcomes
  • Saves hospital resources
  • Includes the following: sterile diversion tube (red top), blood culture bottle adaptor, prevantics antiseptic, sterile gloves, tourniquet, alcohol pads, bandage, tracking decals, and instruction card
Specifications

Specifications

BacTAlert Blood Culture Bottles, BacTAlert Blood Culture Bottles
40/Cs, 40/Cs
Blood, Blood
5 to 25°C, 5 to 25°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit