Learn More
NOTRAX™ Clean-Step™ Tacky Mat
Clean-Step™ Tacky Mats trap and keep out dust and particles
Manufacturer:  NOTRAX™ 080S2436WH
 View more versions of this product
Catalog No. 19-085-981
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Adhesive surface grabs and holds soil matter with no transfer
  • Peel away sheets for fresh, clean surface
  • Numbered tabs ensure one sheet at a time removal and indicate number of remaining layers
  • Sticky mat is not damaged by footwear or wheels, does not slip, is noncontaminating
  • Blue and grey are B level tack whereas white is A level, high level of tack
Specifications
Specifications
24 in.
30 Sheets/Mat, 4 Mats/Cs.
White
36 in.
A Level High Tack