Learn More
Essendant CLEANERCNT and CIRC BD
Supplier:  Essendant AMR1002285
Encompass_Preferred
Safe for use on plastics rubber and metal Fast dry flammable electronics cleaner Must be used only on de-energized components and equipment Non-ozone-depleting Application- Contact Cleaner Applicable Material- Metal Plastic Rubber Dirt Types- Adhesives Dust Grease Grime Oil-Based Soil Water-Based Soil Scent- Unscented
Catalog No. 50-000-22811
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.