Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cleaning Up Oil Spills Naturally
SCI_ED

Simulate an oil spill in water and treat it with a microbial mix to observe the physical changes and degradation of oil film.

Specifications

For Use With (Application) To observe physical changes and degradation of oil film
Includes Ready to use bacterial mixture, oil, petri dishes, vials, pipets, measuring spoon, stirring sticks, sand, gravel, cobbles, gloves, magnifiers and comprehensive teacher and student guides
View More Specs

Requires: Compound microscope


Products 2
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
S23197
SCI_ED
View Documents
Neo/Sci Corporation
201255
Refill Clean Spills This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
S23196
SCI_ED
View Documents
Neo/Sci Corporation
201253
Cleaning Up Oil Spills This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

  • Three spill scenarios: beach sand, gravel and cobbles
  • Observe differences in degradation rates
  • Explore bioremediation methods for other waste chemicals
  • Compare their effectiveness to traditional cleanup methods

Order Info

Class size: 40 students.

Specifications

Specifications

To observe physical changes and degradation of oil film
Ready to use bacterial mixture, oil, petri dishes, vials, pipets, measuring spoon, stirring sticks, sand, gravel, cobbles, gloves, magnifiers and comprehensive teacher and student guides
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit