Brady™ CleanLift™ IDXPERT™ Labels, Repositionable Vinyl Cloth Wire and Cable Wraps (B-498)

Printable, repositionable labels for IDXPERT printers. Brady™ General Identification Labels are made of vinyl film with repositionable rubber based adhesive.

Manufacturer: Brady™ X163498

 View more versions of this product

Catalog No. 19-108-964

  Check Availability
Add to Cart

    Specifications

    Description Compatible surfaces: Powder coated; Rough, smooth, textured
    For Use With (Equipment) IDXPERT printer
    Material B-498 Repositionable vinyl cloth
    Adhesive Type Repositionable rubber
    Wire Gauge 12 to 10 gauge
    Format Cartridge
    For Use With (Application) Barcode Labeling; Cable and Wire Marking; Component and Equipment ID; General and Industrial ID
    Label Size (English) W x H: 0.5 x 1 in.
    Item Description Abrasion, Chemical, Humidity, Corrosion, UV and Weather-Resistant
    Product Type IDXPERT Label
    Quantity 225/Cartridge
    Color Black on white
    Thickness (English) 0.0073 in.
    Certifications/Compliance UL Recognized

    • Narrow-width markers automatically set to fixed spacing and separation between leg ends
    • Rotate to top-down orientation for perfect alignment on terminal blocks
    • Made of repositionable vinyl cloth (B-498)

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit