Learn More
CritiCore Protective Wear Cleanroom Boot
These reusable boots are suitable for a variety of cleanroom applications including: pharmaceutical, biotechnology, and medical device manufacturing.
Supplier: CritiCore Protective Wear 14M12141ESD013XLWHT
This item is not returnable.
View return policy
Description
- High-density, ESD fabric for electrostatic management
- Elastic at Ankle
- Single Strap
- Gamma sterilization stable gray molded sole
- Color-coded strap for easy size identification
- Laser-cut, pre-serged seams
- Silicon-free
- One reinforced snap on back of boot for coverall leg attachment
Specifications
Single-strap Tunneled Elastic with Snaps | |
High Density ESD Polyester | |
Proprietary Molded Sole | |
Non-sterile |
White | |
3X-Large | |
Reusable | |
Yes |