Learn More
Akron Biotechnology LLCSupplier Diversity Partner CLEANSOLUTIONS INCUBTR SPRY 1L DFS Item
Manufacturer:  Akron Biotechnology LLCSupplier Diversity Partner AK52301000
CleanSolutions-Spray is our disinfectant solution that is used for all laboratory surfaces, including incubators and workstations. It prevents the contamination and growth of bacteria, mycoplasma, fungi and viruses.
Catalog No. NC0035289