Learn More
PIP™ CleanTeam™ 12 in. Class 100 Cleanroom Nitrile Gloves
Single-use, finger textured nitrile gloves are recommended for Class 100/ISO 5 cleanrooms and are commonly used in biotechnology, medical device manufacturing and computer assembly.
Specifications
Certifications/Compliance ASTM D5712-05, ASTM D6124-01, IEST-RP-CC005.3
Cleanroom Class Class 100/ISO 5
AQL 4
Thickness (English) 7 mil
Thickness (Metric) 0.18 mm
View More Specs
Products 4
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-121-730A
View Documents
PIP™
100333000/S
Small
Case of 1000 for $251.10
N/A
Due to product restrictions, please to purchase or view availability for this product.
19-121-730B
View Documents
PIP™
100333000/M
Medium
Case of 1000 for $251.10
N/A
Due to product restrictions, please to purchase or view availability for this product.
19-121-730C
View Documents
PIP™
100333000/L
Large
Case of 1000 for $251.10
N/A
Due to product restrictions, please to purchase or view availability for this product.
19-121-730D
View Documents
PIP™
100333000/XL
X-Large
Case of 1000 for $251.10
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
  • Exceed standards required for use in class 100 and other controlled environments
  • Finger textured provides secure grip in wet or dry environments
  • Ambidextrous for quick and easy donning
  • Powder free to reduce allergic reactions
  • Beaded cuff adds additional strength to cuff edge for longer glove life and prevents liquid run-off
  • Chlorinated to harden and cleanse the surface of the glove
  • Electrostatic Dissipative (ESD) for improved performance in electronics assembly
  • 100% Silicone free
Specifications
Specifications
ASTM D5712-05, ASTM D6124-01, IEST-RP-CC005.3
4
0.18 mm
30.5 cm
14 MPa
Class 100/ISO 5
7 mil
12 in.
Fully Textured