Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ CleanTeam™ Economy Nylon Two-Piece Men's Inspection Gloves

Gloves are constructed of low lint 40-denier nylon tricot fabric to absorb perspiration and feature a two-piece pattern with set-in thumb.

Specifications

Color White
Gender Men's
Grade Economy
Hand Ambidextrous
Length (English) 9 in.
View More Specs

Products 1
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-148-385
View Documents
PIP™
98742/L
Large This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

  • Manufacturer-suggested ISO Class 7 (Class 10,000)
  • Economical choice for hand protection or glove liners
  • Made of low-lint, 40-denier tricot fabric that absorbs perspiration for a comfortable fit
  • Two-piece pattern with set-in thumb
  • Color-coded hem for quick size identification
  • Ambidextrous
Specifications

Specifications

White
Economy
9 in.
Inspection Gloves
Keystone
Men's
Ambidextrous
Nylon
98-742
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit