Learn More
CleanWear™ Tyvek™ Disposable Coveralls with Elastic Ankles and Wrists
Tough coverall protects skin from wet and dry particulate matter
Manufacturer:  Cleanwear USA 11542XL
 View more versions of this product
Catalog No. 18-892-405
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Zipper front
Tyvek™ Spunbond Olefin
Yes
Elastic
White
Excellent durability, breathability and barrier protection and ensures wearer comfort by allowing air and moisture vapor to pass through while maintaining a virtually impermeable particulate barrier
Elastic wrist and ankle, collar
Elastic
Collared
Serged
2X-Large