CleanWear™ Tyvek™ Disposable Coveralls with Elastic Ankles and Wrists
Tough coverall protects skin from wet and dry particulate matter
Manufacturer: Cleanwear USA 11544XL
 View more versions of this product
Catalog No. 18-892-407
  Check Availability
Add to Cart
    Specifications
    Material Tyvek™ Spunbond Olefin
    Seam Type Serged
    Color White
    Disposable Yes
    For Use With (Application) Excellent durability, breathability and barrier protection and ensures wearer comfort by allowing air and moisture vapor to pass through while maintaining a virtually impermeable particulate barrier
    Closure Type Zipper front
    Includes Elastic wrist and ankle, collar
    Size 4X-Large
  • Description & Specifications
Related Products
Product Content Feedback