CleanWear™ Tyvek™ Disposable Coveralls with Elastic Ankles and Wrists
Tough coverall protects skin from wet and dry particulate matter
Manufacturer: Cleanwear USA 11545XL
 View more versions of this product
Catalog No. 18-892-408
  Check Availability
Add to Cart
    Specifications
    Material Tyvek™ Spunbond Olefin
    Includes Elastic wrist and ankle, collar
    Seam Type Serged
    Color White
    Size 5X-Large
    Disposable Yes
    For Use With (Application) Excellent durability, breathability and barrier protection and ensures wearer comfort by allowing air and moisture vapor to pass through while maintaining a virtually impermeable particulate barrier
    Closure Type Zipper front
  • Description & Specifications
Related Products
Product Content Feedback